Assay Design



Gene Transcripts

Gene Symbol
PDIL5-2
Entrez Gene ID
840461
Gene Name
PDI-like 5-2
Gene Aliases
ARABIDOPSIS THALIANA PROTEIN DISULFIDE ISOMERASE 8, AT1G35620, ATPDI8, ATPDIL5-2, F15O4.20, PDI-like 5-2, PDI8, PROTEIN DISULFIDE ISOMERASE 8
Chromosome Location
Chr.1: 13156346 - 13158467 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103262.5 NP_564462.1 4 - 5 1151 67
GenBank mRNA AF386986.1 - 4 - 5 1025 67
BT008751.1 - 4 - 5 993 67

View Full Product Details