Assay Design



Gene Transcripts

Gene Symbol
CDPK2
Entrez Gene ID
840471
Gene Name
calcium-dependent protein kinase 2
Gene Aliases
AT1G35670, ATCDPK2, ATCPK11, CALCIUM-DEPENDENT PROTEIN KINASE 2, CPK11, F15O4.8, calcium-dependent protein kinase 2
Chromosome Location
Chr.1: 13205345 - 13208289 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103271.4 NP_174807.1 4 - 5 1063 111
GenBank mRNA AY050981.1 - 4 - 5 1027 111
AY113986.1 - 4 - 5 952 111

View Full Product Details