Assay Design
Gene Transcripts
- Gene Symbol
- TOC75-I
- Entrez Gene ID
-
840488
- Gene Name
- translocon outer membrane complex 75-I
- Gene Aliases
-
AT1G35860, F10O5.4, F10O5_4, translocon outer membrane complex 75-I
- Chromosome Location
-
Chr.1: 13332047 - 13334050 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103285.1 |
NP_174821.1 |
5 - 6 |
782 |
66 |
View Full Product Details