Assay Design
Gene Transcripts
- Gene Symbol
- ACC2
- Entrez Gene ID
-
840522
- Gene Name
- acetyl-CoA carboxylase 2
- Gene Aliases
-
AT1G36180, F15C21.2, F15C21_2, acetyl-CoA carboxylase 2
- Chromosome Location
-
Chr.1: 13545965 - 13558600 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103314.5 |
NP_174850.4 |
23 - 24 |
3233 |
100 |
View Full Product Details