Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001084206.1 | NP_001077675.1 | 2 - 3 | 252 | 127 |
NM_001123963.1 | NP_001117435.1 | 1 - 2 | 258 | 127 | |
NM_103516.4 | NP_175056.1 | 2 - 3 | 288 | 127 | |
GenBank mRNA | AF370263.1 | - | 2 - 3 | 212 | 127 |
AK227664.1 | - | 2 - 3 | 247 | 127 | |
AY063080.1 | - | 1 - 2 | 141 | 127 | |
AY086878.1 | - | 2 - 3 | 251 | 127 | |
U75603.1 | - | 1 - 2 | 160 | 127 |