Assay Design



Gene Transcripts

Gene Symbol
RAB18
Entrez Gene ID
840987
Gene Name
RAB GTPASE HOMOLOG B18
Gene Aliases
ARABIDOPSIS RAB GTPASE HOMOLOG B18, ARABIDOPSIS RAB GTPASE HOMOLOG C1, AT1G43890, ATRAB-C1, ATRAB18, ATRABC1, F28H19.15, F28H19_15, RAB GTPASE HOMOLOG 18-1, RAB GTPASE HOMOLOG B18, RAB GTPASE HOMOLOG C1, RAB18-1, RABC1
Chromosome Location
Chr.1: 16646696 - 16648648 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084206.1 NP_001077675.1 5 - 6 501 121
NM_001123963.1 NP_001117435.1 4 - 5 507 121
NM_103516.4 NP_175056.1 5 - 6 537 121
GenBank mRNA AF370263.1 - 5 - 6 461 121
AK227664.1 - 5 - 6 496 121
AY063080.1 - 4 - 5 390 121
AY086878.1 - 5 - 6 500 121
U75603.1 - 4 - 5 409 121

View Full Product Details