Assay Design
Gene Transcripts
- Gene Symbol
- GA20OX5
- Entrez Gene ID
-
841012
- Gene Name
- gibberellin 20-oxidase 5
- Gene Aliases
-
ARABIDOPSIS THALIANA GIBBERELLIN 20-OXIDASE 5, AT1G44090, ATGA20OX5, T7O23.20, T7O23_20, gibberellin 20-oxidase 5
- Chromosome Location
-
Chr.1: 16760616 - 16762486 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103535.2 |
NP_175075.1 |
2 - 3 |
880 |
72 |
GenBank mRNA |
DQ056484.1 |
- |
2 - 3 |
880 |
72 |
View Full Product Details