Assay Design



Gene Transcripts

Gene Symbol
WOX4
Entrez Gene ID
841113
Gene Name
WUSCHEL related homeobox 4
Gene Aliases
AT1G46480, F2G19.11, F2G19_11, WUSCHEL related homeobox 4
Chromosome Location
Chr.1: 17236524 - 17238185 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103605.7 NP_175145.2 2 - 3 794 109
GenBank mRNA AB493501.1 - 2 - 3 562 109
AY251396.1 - 2 - 3 562 109
BT010692.1 - 2 - 3 662 109
BT010974.1 - 2 - 3 562 109
FJ440850.1 - 2 - 3 595 109

View Full Product Details