Assay Design



Gene Transcripts

Gene Symbol
OSB1
Entrez Gene ID
841183
Gene Name
Primosome PriB/single-strand DNA-binding protein
Gene Aliases
AT1G47720, Organellar Single-stranded, T2E6.21
Chromosome Location
Chr.1: 17561286 - 17562934 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103665.3 NP_175203.2 3 - 4 594 117
GenBank mRNA AK119114.1 - 3 - 4 543 117
BT005269.1 - 3 - 4 519 117

View Full Product Details