Assay Design



Gene Transcripts

Gene Symbol
NPSN12
Entrez Gene ID
841244
Gene Name
Putative plant snare 12
Gene Aliases
AT1G48240, ATNPSN12, F21D18.4, novel plant snare 12
Chromosome Location
Chr.1: 17808988 - 17812106 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333315.1 NP_001323414.1 1 - 2 754 78
NM_103721.5 NP_175258.2 2 - 3 613 78
GenBank mRNA AF487545.1 - 2 - 3 186 78
AK117322.1 - 2 - 3 402 78
BT005271.1 - 2 - 3 163 78

View Full Product Details