Assay Design



Gene Transcripts

Gene Symbol
BS14A
Entrez Gene ID
824986
Gene Name
BET1P/SFT1P-like protein 14A
Gene Aliases
ARABIDOPSIS THALIANA BET1P/SFT1P-LIKE PROTEIN 14A, AT3G58170, ATBET11, ATBS14A, BET11, BET1P/SFT1P-like protein 14A
Chromosome Location
Chr.3: 21542359 - 21544084 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115679.4 NP_191376.1 3 - 4 531 67
GenBank mRNA AF368175.1 - 3 - 4 317 67
AK117351.1 - 3 - 4 388 67
BT004705.1 - 2 - 3 222 67

View Full Product Details