Assay Design



Gene Transcripts

Gene Symbol
CSY2
Entrez Gene ID
825044
Gene Name
citrate synthase 2
Gene Aliases
AT3G58750, citrate synthase 2
Chromosome Location
Chr.3: 21724306 - 21728137 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115737.3 NP_191434.1 4 - 5 1298 111
GenBank mRNA BT015884.1 - 4 - 5 626 111
BT020195.1 - 4 - 5 619 111

View Full Product Details