Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001203206.1 | NP_001190135.1 | 25 - 26 | 3752 | 73 |
NM_001339969.1 | NP_001326041.1 | 25 - 26 | 4619 | 73 | |
NM_001339970.1 | NP_001326042.1 | 24 - 25 | 3761 | 73 | |
NM_001339971.1 | NP_001326040.1 | 24 - 25 | 3532 | 73 | |
NM_115803.3 | NP_191500.2 | 24 - 25 | 3651 | 73 | |
GenBank mRNA | AJ459823.1 | - | 24 - 25 | 3299 | 73 |