Assay Design



Gene Transcripts

Gene Symbol
PBF1
Entrez Gene ID
825253
Gene Name
N-terminal nucleophile aminohydrolases (Ntn hydrolases) superfamily protein
Gene Aliases
20S PROTEASOME BETA SUBUNIT PBF1, AT3G60820
Chromosome Location
Chr.3: 22471769 - 22473971 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035828.2 NP_001030905.1 4 - 5 495 67
NM_001203217.1 NP_001190146.1 4 - 5 527 67
NM_115946.3 NP_191641.1 4 - 5 495 67
GenBank mRNA AF043537.1 - 4 - 5 450 67
AY062790.1 - 4 - 5 464 67
AY081571.1 - 4 - 5 365 67
AY086619.1 - 4 - 5 485 67
X79806.1 - - 424 67

View Full Product Details