Assay Design



Gene Transcripts

Gene Symbol
GAUT1
Entrez Gene ID
825285
Gene Name
galacturonosyltransferase 1
Gene Aliases
AT3G61130, JS36, LGT1, galacturonosyltransferase 1
Chromosome Location
Chr.3: 22621912 - 22625808 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115977.4 NP_191672.1 2 - 3 546 78
GenBank mRNA DQ370437.1 - 1 - 2 144 78
KJ138782.1 - 1 - 2 144 78

View Full Product Details