Assay Design



Gene Transcripts

Gene Symbol
FUS6
Entrez Gene ID
825286
Gene Name
COP9 signalosome complex subunit 1
Gene Aliases
ARABIDOPSIS THALIANA FUSCA 6, AT3G61140, ATFUS6, ATSK31, AtCSN1, CONSTITUTIVE PHOTOMORPHOGENIC 11, COP11, COP9 SIGNALOSOME SUBUNIT 1, CSN1, EMB78, EMBRYO DEFECTIVE 78, FUSCA 6, SHAGGY-LIKE KINASE 31, SK31
Chromosome Location
Chr.3: 22626167 - 22630141 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115978.4 NP_567109.1 4 - 5 1085 90
GenBank mRNA AF360295.1 - 4 - 5 971 90
AF395057.1 - 4 - 5 917 90
AY051056.1 - 4 - 5 917 90

View Full Product Details