Assay Design



Gene Transcripts

Gene Symbol
VSR2
Entrez Gene ID
817579
Gene Name
VACUOLAR SORTING RECEPTOR 2
Gene Aliases
AT2G30290, AtVSR2, BP80-1;2, T9D9.10, T9D9_10, VACUOLAR SORTING RECEPTOR 1;2, VACUOLAR SORTING RECEPTOR 2, VSR1;2, binding protein of 80 kDa 1;2
Chromosome Location
Chr.2: 12912519 - 12915915 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202708.1 NP_001189637.1 1 - 2 430 65
NM_128582.2 NP_180588.1 1 - 2 433 65
GenBank mRNA DQ446580.1 - 1 - 2 299 65

View Full Product Details