Assay Design



Gene Transcripts

Gene Symbol
GSL12
Entrez Gene ID
831140
Gene Name
glucan synthase-like 12
Gene Aliases
AT5G13000, ATGSL12, CALS3, T24H18.170, T24H18_170, callose synthase 3, glucan synthase-like 12, gsl12
Chromosome Location
Chr.5: 4110171 - 4121489 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161240.2 NP_001154712.2 24 - 25 2740 81
NM_121303.8 NP_196804.6 24 - 25 2802 81

View Full Product Details