Assay Design



Gene Transcripts

Gene Symbol
KEG
Entrez Gene ID
831197
Gene Name
protein kinases;ubiquitin-protein ligase
Gene Aliases
AT5G13530, KEEP ON GOING, T6I14.60, T6I14_60
Chromosome Location
Chr.5: 4345202 - 4354723 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203374.1 NP_001190303.1 5 - 6 1880 79
NM_121356.3 NP_196857.2 5 - 6 2204 79
GenBank mRNA DQ315360.1 - 5 - 6 1788 79

View Full Product Details