Assay Design



Gene Transcripts

Gene Symbol
AGL15
Entrez Gene ID
831224
Gene Name
AGAMOUS-like 15
Gene Aliases
AGAMOUS-like 15, AT5G13790, MADS-BOX PROTEIN AGL15, MXE10.8, MXE10_8
Chromosome Location
Chr.5: 4448862 - 4450958 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343293.1 NP_001330207.1 3 - 4 471 119
NM_121382.3 NP_196883.1 3 - 4 368 119

View Full Product Details