Assay Design



Gene Transcripts

Gene Symbol
EXO70C2
Entrez Gene ID
831247
Gene Name
exocyst subunit exo70 family protein C2
Gene Aliases
AT5G13990, ATEXO70C2, MAC12.17, MAC12_17, exocyst subunit exo70 family protein C2
Chromosome Location
Chr.5: 4514568 - 4516892 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121402.2 NP_196903.1 1 - 1 2131 62
GenBank mRNA BT003863.1 - 1 - 1 2131 62
BT006148.1 - 1 - 1 2006 62

View Full Product Details