Assay Design



Gene Transcripts

Gene Symbol
PHT3;1
Entrez Gene ID
831252
Gene Name
phosphate transporter 3;1
Gene Aliases
AT5G14040, MPT3, MUA22.4, MUA22_4, mitochondrial phosphate transporter 3, phosphate transporter 3;1
Chromosome Location
Chr.5: 4530643 - 4533119 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121407.3 NP_196908.1 5 - 6 1222 69
GenBank mRNA AY058848.1 - 5 - 6 1173 69
AY143869.1 - 5 - 6 1068 69

View Full Product Details