Assay Design



Gene Transcripts

Gene Symbol
ABCI11
Entrez Gene ID
831259
Gene Name
non-intrinsic ABC protein 14
Gene Aliases
ARABIDOPSIS THALIANANON-INTRINSIC ABC PROTEIN 14, AT5G14100, ATNAP14, ATP-binding cassette I11, MUA22.10, MUA22_10, NAP14, non-intrinsic ABC protein 14
Chromosome Location
Chr.5: 4549438 - 4551843 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121413.2 NP_196914.1 7 - 8 839 61
GenBank mRNA AK118738.1 - 7 - 8 728 61
AY085446.1 - 7 - 8 839 61
BT006054.1 - 7 - 8 628 61

View Full Product Details