Assay Design
Gene Transcripts
- Gene Symbol
- CRS1
- Entrez Gene ID
-
831476
- Gene Name
- maize chloroplast splicing factor-like protein
- Gene Aliases
-
ARABIDOPSIS ORTHOLOG OF MAIZE CHLOROPLAST SPLICING FACTOR CRS1, AT5G16180, ATCRS1, T21H19.100, T21H19_100, ortholog of maize chloroplast splicing factor CRS1
- Chromosome Location
-
Chr.5: 5279884 - 5283102 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121623.3 |
NP_197122.2 |
4 - 5 |
1622 |
106 |
View Full Product Details