Assay Design
Gene Transcripts
- Gene Symbol
- BGLU2
- Entrez Gene ID
-
831520
- Gene Name
- beta glucosidase 2
- Gene Aliases
-
AT5G16580, MTG13.2, MTG13_2, beta glucosidase 2
- Chromosome Location
-
Chr.5: 5425641 - 5427472 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121664.3 |
NP_197161.2 |
3 - 4 |
432 |
92 |
View Full Product Details