Assay Design



Gene Transcripts

Gene Symbol
TIC40
Entrez Gene ID
831524
Gene Name
hydroxyproline-rich glycoprotein family protein
Gene Aliases
AT5G16620, ATTIC40, MTG13.14, MTG13_14, PDE120, pigment defective embryo, pigment defective embryo 120, translocon at the inner envelope membrane of chloroplasts 40
Chromosome Location
Chr.5: 5450620 - 5454577 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121668.4 NP_197165.1 11 - 12 1325 130
GenBank mRNA AF428299.1 - 11 - 12 1247 130
AY093010.1 - 11 - 12 1208 130
BT006595.1 - 11 - 12 1137 130

View Full Product Details