Assay Design



Gene Transcripts

Gene Symbol
RAD4
Entrez Gene ID
831525
Gene Name
DNA repair protein Rad4 family
Gene Aliases
AT5G16630, ATRAD4, MTG13.7, MTG13_7
Chromosome Location
Chr.5: 5454654 - 5459647 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036817.3 NP_001031894.1 7 - 8 1038 82
NM_001343458.1 NP_001332636.1 7 - 8 1038 82
NM_121669.3 NP_197166.2 7 - 8 913 82
GenBank mRNA AY062755.1 - 7 - 8 886 82
BT010359.1 - 6 - 7 743 82

View Full Product Details