Assay Design



Gene Transcripts

Gene Symbol
RAD4
Entrez Gene ID
831525
Gene Name
DNA repair protein Rad4 family
Gene Aliases
AT5G16630, ATRAD4, MTG13.7, MTG13_7
Chromosome Location
Chr.5: 5454654 - 5459647 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036817.3 NP_001031894.1 9 - 10 1953 85
NM_001343458.1 NP_001332636.1 9 - 10 1953 85
NM_121669.3 NP_197166.2 9 - 10 1828 85
GenBank mRNA AY062755.1 - 9 - 10 1801 85
BT010359.1 - 8 - 9 1658 85

View Full Product Details