Assay Design



Gene Transcripts

Gene Symbol
APG5
Entrez Gene ID
831594
Gene Name
autophagy protein Apg5 family
Gene Aliases
AT5G17290, ATATG5, ATG5, AUTOPHAGY 5, MKP11.20, MKP11_20
Chromosome Location
Chr.5: 5686135 - 5689947 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121735.5 NP_197231.1 4 - 5 514 123
GenBank mRNA AK117572.1 - 4 - 5 400 123
BT005100.1 - 4 - 5 388 123

View Full Product Details