Assay Design
Gene Transcripts
- Gene Symbol
- HDG9
- Entrez Gene ID
-
831598
- Gene Name
- homeodomain GLABROUS 9
- Gene Aliases
-
AT5G17320, MKP11.28, MKP11_28, homeodomain GLABROUS 9
- Chromosome Location
-
Chr.5: 5703139 - 5707637 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121738.2 |
NP_197234.1 |
4 - 5 |
935 |
91 |
View Full Product Details