Assay Design



Gene Transcripts

Gene Symbol
CBR
Entrez Gene ID
831645
Gene Name
NADH:cytochrome B5 reductase 1
Gene Aliases
AT5G17770, ATCBR, CBR1, NADH:CYTOCHROME B5 REDUCTASE 1, NADH:cytochrome B5 reductase 1
Chromosome Location
Chr.5: 5864182 - 5866809 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121783.5 NP_197279.1 5 - 6 825 74
GenBank mRNA AB007799.1 - 5 - 6 593 74
AK316787.1 - 5 - 6 536 74
AY085728.1 - 5 - 6 664 74

View Full Product Details