Assay Design



Gene Transcripts

Gene Symbol
APY2
Entrez Gene ID
831946
Gene Name
apyrase 2
Gene Aliases
APYRASE, AT5G18280, ATAPY2, MRG7.24, MRG7_24, apyrase 2
Chromosome Location
Chr.5: 6050514 - 6054386 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121833.7 NP_197329.5 7 - 8 1475 90
GenBank mRNA AF156783.1 - 7 - 8 1430 90
AY099866.1 - 7 - 8 1421 90
BT000329.1 - 7 - 8 1112 90

View Full Product Details