Assay Design
Gene Transcripts
- Gene Symbol
- SAD2
- Entrez Gene ID
-
817722
- Gene Name
- ARM repeat superfamily protein
- Gene Aliases
-
AT2G31660, EMA1, SUPER SENSITIVE TO ABA AND DROUGHT2, T9H9.18, T9H9_18, UNARMED 9, URM9, enhanced miRNA activity 1
- Chromosome Location
-
Chr.2: 13464294 - 13471867 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128723.3 |
NP_180724.2 |
7 - 8 |
1220 |
95 |
View Full Product Details