Assay Design



Gene Transcripts

Gene Symbol
CHX21
Entrez Gene ID
817749
Gene Name
cation/H+ exchanger 21
Gene Aliases
AT2G31910, ATCHX21, F20M17.5, F20M17_5, cation/H+ exchanger 21
Chromosome Location
Chr.2: 13570107 - 13574632 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336355.1 NP_001324589.1 3 - 4 692 113
NM_001336356.1 NP_001324588.1 2 - 3 483 113
NM_128749.2 NP_180750.2 2 - 3 672 113

View Full Product Details