Assay Design
Gene Transcripts
- Gene Symbol
- HDG3
- Entrez Gene ID
-
817798
- Gene Name
- homeodomain GLABROUS 3
- Gene Aliases
-
AT2G32370, T32F6.11, T32F6_11, homeodomain GLABROUS 3
- Chromosome Location
-
Chr.2: 13740878 - 13746286 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128796.3 |
NP_180796.2 |
5 - 6 |
801 |
69 |
GenBank mRNA |
EF988635.1 |
- |
5 - 6 |
801 |
69 |
View Full Product Details