Assay Design



Gene Transcripts

Gene Symbol
RLP22
Entrez Gene ID
817826
Gene Name
receptor like protein 22
Gene Aliases
AT2G32660, AtRLP22, F24L7.20, F24L7_20, receptor like protein 22
Chromosome Location
Chr.2: 13853699 - 13856297 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336409.1 NP_001323933.1 1 - 1 1721 101
NM_128825.3 NP_180825.2 1 - 1 1536 101
GenBank mRNA AK228145.1 - 1 - 1 1536 101

View Full Product Details