Assay Design
Gene Transcripts
- Gene Symbol
- AGO6
- Entrez Gene ID
-
817856
- Gene Name
- Argonaute family protein
- Gene Aliases
-
ARGONAUTE 6, AT2G32940, T21L14.12, T21L14_12
- Chromosome Location
-
Chr.2: 13971916 - 13977521 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128854.4 |
NP_180853.2 |
15 - 16 |
2101 |
62 |
View Full Product Details