Assay Design
Gene Transcripts
- Gene Symbol
- RLP27
- Entrez Gene ID
-
817868
- Gene Name
- receptor like protein 27
- Gene Aliases
-
AT2G33060, AtRLP27, F25I18.20, F25I18_20, receptor like protein 27
- Chromosome Location
-
Chr.2: 14025483 - 14028261 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128866.4 |
NP_180865.2 |
1 - 1 |
1336 |
104 |
View Full Product Details