Assay Design



Gene Transcripts

Gene Symbol
NUDT22
Entrez Gene ID
817959
Gene Name
nudix hydrolase homolog 22
Gene Aliases
AT2G33980, T14G11.10, T14G11_10, atnudt22, nudix hydrolase homolog 22
Chromosome Location
Chr.2: 14349764 - 14360058 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336478.1 NP_001325146.1 4 - 5 1002 86
NM_128949.3 NP_565776.1 4 - 5 845 86
GenBank mRNA AF360290.1 - 4 - 5 917 86
AK175540.1 - 4 - 5 843 86
AY051046.1 - 4 - 5 784 86

View Full Product Details