Assay Design
Gene Transcripts
- Gene Symbol
- NUDT22
- Entrez Gene ID
-
817959
- Gene Name
- nudix hydrolase homolog 22
- Gene Aliases
-
AT2G33980, T14G11.10, T14G11_10, atnudt22, nudix hydrolase homolog 22
- Chromosome Location
-
Chr.2: 14349764 - 14360058 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128949.3 |
NP_565776.1 |
7 - 8 |
1105 |
91 |
GenBank mRNA |
AF360290.1 |
- |
7 - 8 |
1177 |
91 |
View Full Product Details