Assay Design
Gene Transcripts
- Gene Symbol
- iqd9
- Entrez Gene ID
-
817960
- Gene Name
- IQ-domain 9
- Gene Aliases
-
AT2G33990, IQ-domain 9, T14G11.11, T14G11_11
- Chromosome Location
-
Chr.2: 14360255 - 14362511 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128950.5 |
NP_180946.2 |
2 - 3 |
537 |
116 |
GenBank mRNA |
AK229389.1 |
- |
2 - 3 |
204 |
116 |
View Full Product Details