Assay Design



Gene Transcripts

Gene Symbol
NFU5
Entrez Gene ID
841563
Gene Name
NFU domain protein 5
Gene Aliases
AT1G51390, ATNFU1, F11M15.25, F11M15_25, NFU domain protein 5
Chromosome Location
Chr.1: 19050298 - 19052094 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104017.4 NP_175550.2 2 - 3 503 143
GenBank mRNA AJ512937.1 - 2 - 3 404 143
AK226520.1 - 2 - 3 458 143
BT025642.1 - 2 - 3 374 143

View Full Product Details