Assay Design
Gene Transcripts
- Gene Symbol
- EXO70G2
- Entrez Gene ID
-
841589
- Gene Name
- exocyst subunit exo70 family protein G2
- Gene Aliases
-
AT1G51640, ATEXO70G2, F19C24.13, F19C24_13, exocyst subunit exo70 family protein G2
- Chromosome Location
-
Chr.1: 19149761 - 19152288 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104042.2 |
NP_175575.1 |
1 - 1 |
1970 |
99 |
View Full Product Details