Assay Design



Gene Transcripts

Gene Symbol
B ALPHA
Entrez Gene ID
841594
Gene Name
protein phosphatase 2A 55 kDa regulatory subunit B alpha
Gene Aliases
AT1G51690, ATB ALPHA, F19C24.10, F19C24_10, protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform
Chromosome Location
Chr.1: 19163843 - 19170449 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198270.1 NP_001185199.1 15 - 16 1731 90
NM_001333494.1 NP_001322512.1 13 - 14 3630 90
NM_001333495.1 NP_001322511.1 15 - 16 1752 90
NM_104047.4 NP_564595.1 14 - 15 3359 90
NM_202274.2 NP_974003.1 13 - 14 1653 90
GenBank mRNA AK230310.1 - 14 - 15 3139 90
AY120756.1 - 13 - 14 1654 90
BT000109.1 - 13 - 14 1412 90

View Full Product Details