Assay Design



Gene Transcripts

Gene Symbol
CYSC1
Entrez Gene ID
825317
Gene Name
cysteine synthase C1
Gene Aliases
ARATH;BSAS3;1, AT3G61440, ATCYSC1, BETA-SUBSTITUTED ALA SYNTHASE 3;1, CYSTEINE SYNTHASE, CYSTEINE SYNTHASE C1, cysteine synthase C1
Chromosome Location
Chr.3: 22735663 - 22738131 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084855.2 NP_001078324.1 3 - 4 554 75
NM_001203223.1 NP_001190152.1 3 - 4 756 75
NM_001340099.1 NP_001327883.1 4 - 5 730 75
NM_116009.4 NP_191703.1 4 - 5 798 75
GenBank mRNA AB024282.1 - 4 - 5 646 75
AJ010505.1 - 4 - 5 675 75
AK226606.1 - 4 - 5 668 75
AY087208.1 - 4 - 5 730 75
AY093094.1 - 4 - 5 732 75
AY128782.1 - 4 - 5 592 75

View Full Product Details