Assay Design



Gene Transcripts

Gene Symbol
RRP41
Entrez Gene ID
825335
Gene Name
3'-5'-exoribonuclease family protein
Gene Aliases
AT3G61620, EXONUCLEASE RRP41
Chromosome Location
Chr.3: 22801215 - 22804104 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125406.1 NP_001118878.1 7 - 8 605 106
NM_116027.5 NP_191721.1 6 - 7 447 106
GenBank mRNA AF191741.1 - 6 - 7 336 106
AK118846.1 - 6 - 7 363 106
BT005485.1 - 5 - 6 265 106

View Full Product Details