Assay Design



Gene Transcripts

Gene Symbol
SDG14
Entrez Gene ID
825347
Gene Name
SET domain protein 14
Gene Aliases
AT3G61740, ATX3, F21F14.13, SET domain protein 14
Chromosome Location
Chr.3: 22850811 - 22857016 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084857.2 NP_001078326.1 2 - 3 1254 100
NM_116039.4 NP_191733.3 2 - 3 1210 100
GenBank mRNA AB493660.1 - 2 - 3 786 100

View Full Product Details