Assay Design



Gene Transcripts

Gene Symbol
BGLU7
Entrez Gene ID
825449
Gene Name
beta glucosidase 7
Gene Aliases
AT3G62740, beta glucosidase 7
Chromosome Location
Chr.3: 23211287 - 23214066 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340171.1 NP_001327167.1 2 - 3 313 141
NM_116139.3 NP_191833.2 2 - 3 313 141

View Full Product Details