Assay Design
Gene Transcripts
- Gene Symbol
- RPS13A
- Entrez Gene ID
-
828167
- Gene Name
- ribosomal protein S13A
- Gene Aliases
-
AT4G00100, ATRPS13A, F6N15.7, F6N15_7, PFL2, POINTED FIRST LEAF 2, RIBOSOMAL PROTEIN S13, RPS13, ribosomal protein S13A
- Chromosome Location
-
Chr.4: 36916 - 38384 on Build TAIR10.1
- Gene Symbol
- FIB1
- Entrez Gene ID
-
835323
- Gene Name
- fibrillarin 1
- Gene Aliases
-
AT5G52470, ATFBR1, ATFIB1, FBR1, FIBRILLARIN 1, K24M7.22, K24M7_22, SKIP7, SKP1/ASK1-INTERACTING PROTEIN, fibrillarin 1
- Chromosome Location
-
Chr.5: 21294147 - 21296819 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK230239.1 |
- |
- |
251 |
92 |
View Full Product Details