Assay Design



Gene Transcripts

Gene Symbol
XSP1
Entrez Gene ID
827949
Gene Name
xylem serine peptidase 1
Gene Aliases
AT4G00230, F6N15.3, F6N15_3, SUBTILISIN-TYPE SERINE ENDOPEPTIDASE XSP1, xylem serine peptidase 1
Chromosome Location
Chr.4: 93743 - 97467 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340236.1 NP_001328541.1 9 - 10 1535 72
NM_116240.5 NP_567155.1 9 - 10 1686 72
GenBank mRNA AF190794.1 - 9 - 10 1506 72

View Full Product Details